Order Entry
Spain
ContactUsLinkComponent
Anti-CAP1 Antibody
Anti-CAP1 Antibody
Catalog # BSBTPB9923
Supplier:  Boster Bio
undefined
Anti-CAP1 Antibody
Catalog # BSBTPB9923
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CAP, adenylate cyclase-associated protein 1 (yeast)
  • Antigen symbol:
    CAP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    10487
  • Antigen synonyms:
    CAP|CAP1-PEN
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Adenylyl cyclase-associated protein 1(CAP1) detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: CAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat