Order Entry
Spain
ContactUsLinkComponent
Test kits, residual hardness cell test, Spectroquant®, Supelco®
Test kits, residual hardness cell test, Spectroquant®, Supelco®
Catalog # 1.14683.0001
Supplier:  Merck
CAS Number:  
undefined
Test kits, residual hardness cell test, Spectroquant®, Supelco®
Catalog # 1.14683.0001
Supplier:  Merck
CAS Number:  
Specifications
  • Chemical method:
    Phthalein complexone
  • Pk:
    25 Tests
  • Description:
    Residual hardness cell test
  • Environmentally Preferable:
  • M-Clarity quality level:
    MQ100
  • For unit:
    Prove, Nova 30/60, Multy, Move
  • Range:
    0,50 - 5,00 mg/l Ca, 0,070 - 0,700 mg/l °d, 0,087 - 0,874 °e, 0,12 - 1,25 °f, 0,70 - 7,00 mg/l CaO, 1,2 - 12,5 CaCO₃
Specifications
About this item
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.