Order Entry
Spain
ContactUsLinkComponent
 
Dimethyl sulphoxide ≥99.5% for LC-MS, Pierce™
Catalog # PIER85190
CAS Number:  
undefined
Dimethyl sulphoxide ≥99.5% for LC-MS, Pierce™
Catalog # PIER85190
CAS Number:  

Specifications

  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • Visual Appearance:
    Clear, Colorless liquid, Free of particulate matter
  • Purity:
    ≥ 99.5%
  • Water Content:
    ≤ 0.2%

Specification Test Results

About this item

 :
 :78.14 g/mol
Boiling Pt:189 °C (1013 hPa)
Melting Pt:18.5 °C
Density:1.101 g/cm³ (20 °C)
Flash Pt:87 °C
CAS Number:67-68-5
Formula:(CH₃)₂SO
MDL Number:MFCD00002089