VWR® Buffer, TAE, 50X for Electrophoresis
Supplier: PROLABO CMP
Certificates
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Serological pipettes
Supplier: Greiner Bio-One
Test kits, phosphate, Spectroquant®, Supelco®
Supplier: Merck
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
VWR®, Strips of PCR Tubes and Caps
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
Sodium hydroxide, pellets, pure, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
PIG® eco-friendly spill response bag
Supplier: NEW PIG
Wipes for delicate tasks, Kimtech™ Science
Supplier: KIMBERLY CLARK