Calcium citrate tetrahydrate, powder USP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:5785-44-4
:
Formula:C₁₂H₁₀Ca₃O₁₄·4H₂O
MDL Number:MFCD00150786
:570.5 g/mol
EINECS:212-391-7
Specification Test Results
- Assay:97.5% Min.
Specification Test Results
Frequently Bought Together