Sodium metabisulphite ≥65%, granular NF, Ph. Eur., BP, JP for food industry
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:7681-57-4
:
Formula:Na₂S₂O₅
MDL Number:MFCD00167602
:190.11 g/mol
Merck Index:12,08784
Melting Pt:150 °C
Density:1.48 g/cm³ (20 °C)
EINECS:231-673-0
REACH:01-2119531326-45
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Assay:65% Min.
Specification Test Results
Frequently Bought Together