Sodium L(+)-ascorbate USP
Catalog # SPCMSO108-50KGBL
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pk:50 kg
- Seq:4418
Specifications
Specification Test Results
- Assay:99% Min.
Specification Test Results
About this item
:
:198.11 g/mol
Melting Pt:219 °C
Density:1.799 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-126-1
CAS Number:134-03-2
Formula:C₆H₇NaO₆
MDL Number:MFCD00082340