Aluminium sulphate hydrate USP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:17927-65-0
:MFCD00149138
Formula:Al₂(SO₄)₃·xH₂O
MDL Number:MFCD00149136
:366.21 g/mol
EINECS:233-135-0
:GHS05
:GHS07
Specification Test Results
- Assay [Al2(SO4)3]:54-59.0 %
Specification Test Results
Frequently Bought Together