Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
p-Anisic acid
p-Anisic acid
p-Anisic acid
p-Anisic acid
SPCMA2287-10KGBL
:
Spectrum Chemical
:
p-Anisic acid
SPCMA2287-10KGBL
:
Spectrum Chemical
:
Pack type:
Poly pail
Pk:
10 kg
Seq:
4158
Assay:
98% Min.
:
:
152.15 g/mol
:
13,00669
:
275 °C (1013 hPa)
:
182…185 °C
:
∼1.385 g/cm³ (20 °C)
:
Ambient
:
202-818-5
:
100-09-4
:
C₈H₈O₃
:
MFCD00002542