Gum arabic, spray-dried powder NF
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:9000-01-5
:
MDL Number:MFCD00081264
Density:1.35 g/cm³ (20 °C)
Flash Pt:>250 °C
EINECS:232-519-5
:GHS07
Storage Temperature:Ambient
Frequently Bought Together