

Cefsulodin sodium salt hydrate 97%
Catalog # J66120.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
Specifications
About this item
:
:554.53 g/mol (anhy)
Merck Index:14,1945
Melting Pt:175 °C
EINECS:257-692-4
:GHS08
:GHS07
Storage Temperature:Freezer
CAS Number:1426397-23-0
Formula:C₂₂H₁₉N₄NaO₈S₂·xH₂O
MDL Number:MFCD07793338