Order Entry
Spain
ContactUsLinkComponent
(R)-1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid 95%
Irritant
(R)-1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid 95%
Catalog # H57825.03
CAS Number:  
Irritant
(R)-1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid 95%
Catalog # H57825.03
CAS Number:  

Specifications

  • Pk:
    1 g

Specifications

About this item

CAS Number:72925-16-7
 :
Formula:C₁₀H₁₇NO₄
MDL Number:MFCD03094727
 :215.25 g/mol
 :GHS07