3,5-Difluoro-N-fmoc-L-phenylalanine 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:205526-24-5
:
Formula:C₂₄H₁₉F₂NO₄
MDL Number:MFCD00797579
:423.4 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together