Order Entry
Spain
Orders LinkContactUsLinkComponent
Ac-β-Ala-OH 97%

Ac-β-Ala-OH 97%

Ac-β-Ala-OH 97%
Certificates
Ac-β-Ala-OH 97%
Ac-β-Ala-OH 97%

2 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 3025-95-4 Molecular Formula: C5H9NO3 Formula Weight: 131.13 Melting Point: 77° MDL No.: MFCD00037296

Synonyms:  N-Ac-β-Ala-OH, 3-Acetamidopropanoic acid, N-Acetyl-beta-alanine, N-Acetyl-b-alanine
  • CAS Number:3025-95-4
  • Formula:C₅H₉NO₃
  • MDL Number:MFCD00037296
  • Molecular Weight:131.1 g/mol
  • Melting Pt:77 °C
  • Storage Temperature:Ambient
  • EINECS:221-185-6
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.