

2-Bromohexanoic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:616-05-7
:
Formula:C₆H₁₁BrO₂
MDL Number:MFCD00004218
:195.06 g/mol
Merck Index:14,1415
Boiling Pt:136…138 °C (18 mmHg)
Melting Pt:3…5 °C
Density:1.37
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:210-461-1
ADR:8,II
:GHS05
:GHS07
Frequently Bought Together