Order Entry
Spain
ContactUsLinkComponent
Sulphur in diesel fuel, standard solutions, Specpure®
Sulphur in diesel fuel, standard solutions, Specpure®
Certificates
Sulphur in diesel fuel, standard solutions, Specpure®
Certificates

Specifications

  • MDL No.:
    MFCD01095233
  • Density:
    0.6-0.9 g/mL
  • Flash point:
    69°C(156°F)
  • Physical form:
    Liquid
  • Boiling point:
    171-315°
  • Appearance:
    Light brown
  • Odour/scent:
    Petroleum-like

Specifications

Frequently Bought Together