Order Entry
Spain
ContactUsLinkComponent
Ion chromatography standard solution, acetate, Specpure®, 1000 µg/ml
Ion chromatography standard solution, acetate, Specpure®, 1000 µg/ml
Certificates
Ion chromatography standard solution, acetate, Specpure®, 1000 µg/ml
Certificates

Specifications

  • MDL No.:
    MFCD00282007
  • Physical form:
    Liquid
  • Odour/scent:
    Odourless
  • Molecular formula:
    CH3CO2Na in H2O

Specifications

Related Information

Frequently Bought Together