Disinfectant tablets
Catalog # ELGACT3
Supplier: ELGA LabWater
Specifications
- Description:Disinfectant tablets
- Seq:02
- Packed:12/pack
- Pk:12
Specifications
About this item
These disinfectant tablets sanitises tubing or pipe work to remove bacteria, reduce bio film and protect the system against bacterial growth.