Surface cleaner, floors, TASKI Jontec Forward
DIVY7513111
: Diversey
- Description:TASKI Jontec Forward, floor cleaner and surface disinfectant,
- Packed:10 litre canister
- Pk:10 L
Highly effective, alkaline floor cleaner, designed for use in scrubber driers.
- Slightly alkaline, low foaming
- High emulsifying power
- Manual or machine application