Order Entry
Spain
ContactUsLinkComponent
Magnetic beads, Sera-Mag Oligo (dT) coated magnetic particles
Magnetic beads, Sera-Mag Oligo (dT) coated magnetic particles
Supplier:  Cytiva
Certificates
undefined
Magnetic beads, Sera-Mag Oligo (dT) coated magnetic particles
Supplier:  Cytiva
Specifications
  • Catalog No:
  • 38152103010350.
  • 38152103011150.
  • 38152103010150.
  • Binding capacity:
  • 11 µg mRNA/mg beads
  • 11 µg mRNA/mg beads
  • 11 µg mRNA/mg beads
  • Particle size:
  • nominal 1 µm diameter
  • nominal 1 µm diameter
  • nominal 1 µm diameter