Order Entry
Spain
ContactUsLinkComponent
Trifluoromethanesulphonic acid 0.1 mol/l in anhydrous acetic acid, Titripur®, Supelco®
Flammable
Corrosive
Trifluoromethanesulphonic acid 0.1 mol/l in anhydrous acetic acid, Titripur®, Supelco®
Supplier:  Merck
Certificates
Flammable
Corrosive
Trifluoromethanesulphonic acid 0.1 mol/l in anhydrous acetic acid, Titripur®, Supelco®
Supplier:  Merck

Specification Test Results

  • Form:
    liquid
  • Amount-of-substance concentration:
    0.0995 - 0.1005
  • Measurement uncertainty:
    +/- 0.0003
  • Traceability:
    NIST SRM
  • The concentration is determined by volumetric titration and refers to 20°C.:
    (20 °C) soluble
  • Storage:
    +15...+25 °C

Specification Test Results

Related Information

Frequently Bought Together