Perchloric acid 69 - 72%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7601-90-3
:
Formula:HClO₄
MDL Number:MFCD00011325
:100.46 g/mol
Boiling Pt:160…181 °C (1013 hPa)
Melting Pt:–112.2 °C
Density:1.53…1.768 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-512-4
ADR:5.1,I
:Management of Change (MOC) category = N
UN:1873
:GHS03
:GHS05
Specification Test Results
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:—
- ACS - Assay (HClO₄):69.0 - 72.0 %
- ACS - Color (APHA):≤ 10
- ACS - Residue after Ignition:≤ 0.003 %
- ACS - Chloride (Cl):≤ 0.001 %
- ACS - Nitrogen Compounds (as N):≤ 0.001 %
- ACS - Sulfate (SO₄):≤ 0.001 %
- ACS - Silicate and Phosphate (as SiO₂):≤ 5 ppm
- ACS - Heavy Metals (by ICP-OES):≤ 1 ppm
- Trace Impurities - Iron (Fe):≤ 1 ppm
Specification Test Results
Frequently Bought Together