4,5-Diamino-6-hydroxy-2-pyrimidinethiol hemisulphate-1,5-hydrate for synthesis, Sigma-Aldrich®
Catalog # 8.14702.0005
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Melting Point:235 °C
- Hill Formula:C8H14N8O6S3*(H2O)n
- Assay:≥97,0% (a/a)
- Identity:Passes test
Specification Test Results
About this item
:
:414.442 g/mol
:M-Clarity™ quality level = MQ200
CAS Number:207226-25-3
Formula:C₈H₁₄N₈O₆S₃