Anti-Prostaglandin E Receptor EP4 Rabbit Polyclonal Antibody
Catalog # BOSSBS-8538R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Prostaglandin E Receptor EP4
- Antigen symbol:PTGER4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Gene ID:5734
- Antigen synonyms:PGE2 receptor EP4 subtype|PGE receptor EP4 subtype|Prostaglandin E2 receptor EP4 subtype|EP4|PTGER2|EP4R|Prostanoid EP4 receptor|PTGER4
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:161-184/488
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Receptor for prostaglandin E2 (PGE2). The activity of this receptor is mediated by G(s) proteins that stimulate adenylate cyclase. Has a relaxing effect on smooth muscle. May play an important role in regulating renal hemodynamics, intestinal epithelial transport, adrenal aldosterone secretion, and uterine function.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: Prostaglandin E Receptor EP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat