Order Entry
Spain
ContactUsLinkComponent
Anti-CTAGE5 Rabbit Polyclonal Antibody
Anti-CTAGE5 Rabbit Polyclonal Antibody
Catalog # BOSSBS-6808R
Supplier:  Bioss
Anti-CTAGE5 Rabbit Polyclonal Antibody
Catalog # BOSSBS-6808R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CTAGE family, member 5
  • Antigen symbol:
    CTAGE5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    4253
  • Antigen synonyms:
    MGEA|MEA6|MGEA11|MGEA6
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Cutaneous T cell lymphomas (CTCL) represent a group of malignancies that originate from CD4-T lymphocytes and manifest on the skin. CTCL is a general term for several neoplasms including mycosis fungoides, T cell leukemia/ lymphoma and pagetoid reticulosis, all of which are very difficult to treat in the advanced stages. CTAGE2 (cutaneous T cell lymphoma associated-antigen 2) is a member of the cancer/testis antigen family of proteins (CTAGE) that, under normal conditions, are found primarily in the testis with little or no expression in other parts of the body. Another member of the CTAG family, namely CTAGE5 (also known as MEA11, MEA6, MGEA11 or MGEA6), is a tumor associated antigen that is found in tumors of various origins, including cutaneous T cell lymphomas. Both CTAGE2 and CTAGE5 are expressed as multiple isoforms due to alternative splicing events.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CTAGE5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: