Order Entry
Spain
ContactUsLinkComponent
Anti-ITM2C Rabbit Polyclonal Antibody
Anti-ITM2C Rabbit Polyclonal Antibody
Catalog # BOSSBS-11859R
Supplier:  Bioss
Anti-ITM2C Rabbit Polyclonal Antibody
Catalog # BOSSBS-11859R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Integral Membrane Protein 2C
  • Antigen symbol:
    ITM2C
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Antigen synonyms:
    E25|BRI3|E25C|BRICD2C|ITM3
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Negative regulator of beta amyloid peptide production. May inhibit the processing of APP by blocking its access to alpha-and beta-secretase. Binding to the beta-secretase-cleaved APP C-terminal fragment is negligible, suggesting that ITM2C is a poor gamma-secretase cleavage inhibitor. May play a role in TNF-induced cell death and neuronal differentiation.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: ITM2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat