Order Entry
Spain
ContactUsLinkComponent
Anti-Puratrophin 1/PLEKHG4 Rabbit Polyclonal Antibody
Anti-Puratrophin 1/PLEKHG4 Rabbit Polyclonal Antibody
Catalog # BOSSBS-11747R
Supplier:  Bioss
Anti-Puratrophin 1/PLEKHG4 Rabbit Polyclonal Antibody
Catalog # BOSSBS-11747R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Puratrophin 1
  • Antigen symbol:
    PLEKHG4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Antigen synonyms:
    ARHGEF44, PH domain-containing family G member 4, PKHG4_HUMAN, Pleckstrin homology domain containing family G with RhoGef domain member 4, member 4, Pleckstrin homology domain-containing family G member 4, PLEKHG 4, PLEKHG4, PRTPHN 1, PRTPHN1, Puratrophin 1, Puratrophin-1, Puratrophin1, Purkinje cell atrophy associated protein 1, Purkinje cell atrophy-associated protein 1, SCA 4, SCA4.
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Possible role in intracellular signaling and cytoskeleton dynamics at the Golgi.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Puratrophin 1/PLEKHG4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat