Order Entry
Spain
ContactUsLinkComponent
Tetraethylenepentamine (mixture of aliphatic amines) for synthesis pH 11.8 (20 g/l, H₂O, 20 °C)., Sigma-Aldrich®
Tetraethylenepentamine (mixture of aliphatic amines) for synthesis pH 11.8 (20 g/l, H₂O, 20 °C)., Sigma-Aldrich®
Supplier:  Merck
Certificates
Tetraethylenepentamine (mixture of aliphatic amines) for synthesis pH 11.8 (20 g/l, H₂O, 20 °C)., Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Assay (GC, area%):
    ≥ 95.0 % (a/a)
  • Water (K. F.):
    ≤ 0.50 %
  • Identity (IR):
    passes test
  • Boiling Point:
    330 °C (1013 hPa)
  • Vapor Pressure:
     - 0.1 hPa (20 °C)
  • Flash Point:
    193 °C
  • Explosion Limit:
    0.1 - 15 %(V)
  • Density:
    0.998 g/cm3 (20 °C)
  • pH:
    11.8 (20 g/l, H2O, 20 °C)
  • Ignition Point:
    330 °C
  • Synonyms:
    1,4,7,10,13-Pentaazatridecane
  • Hill Formula:
    C8H23N5
  • Solubility:
    6540 g/l
  • Storage temperature:
    +15...+25 °C

Specification Test Results

Related Information

Frequently Bought Together