Specifications
- Antibody type:Primary
- Antigen symbol:NBR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Oryza sativa
- Western blot:Yes
- UniProtKB:Q9SB64
- Immunogen:UNA2 domain of NBR1 of Arabidopsis thaliana, fused with GST,
- Size:50 µl
- Pk:50 µl
Specifications
About this item
NBR1 (Autophagy substrate NBR1) is involved in selective autophagy process of damaged organelles, intracellular microbes, protein aggregates, cellular structures and specific soluble proteins. NBR1 mediates the process as an autophagic adapter. The protein has two UBA domains but only the C-terminal UBA domain bound ubiquitin. Alternative names: At4g24690, Putative uncharacterized protein F22K18.110.,NBR1 (Autophagy substrate NBR1) is involved in selective autophagy process of damaged organelles, intracellular microbes, protein aggregates, cellular structures and specific soluble proteins. NBR1 mediates the process as an autophagic adapter. The protein has two UBA domains but only the C-terminal UBA domain bound ubiquitin. Alternative names: At4g24690, Putative uncharacterized protein F22K18.110.
Type: Primary
Antigen: NBR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Oryza sativa