Order Entry
Spain
ContactUsLinkComponent
Anti-HSPA9 Rabbit Polyclonal Antibody
Anti-HSPA9 Rabbit Polyclonal Antibody
  BSBTPB9642
undefined
Anti-HSPA9 Rabbit Polyclonal Antibody
  BSBTPB9642

 

  • Antigen name:
    Heat shock 70kDa protein 9
  • Antigen symbol:
    HSPA9
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Gene ID:
    3313
  • Antigen synonyms:
    MOT|GRP-75|CRP40|GRP75|MOT2|HSPA9B|CSA|HEL-S-124m|PBP74|MTHSP75
  • UniProtKB:
    P38646
  • Storage temperature:
    At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human Grp75 (646-679aa KLFEMAYKKMASEREGSGSSGTGEQKEDQKEEKQ), identical to the related mouse and rat sequences.
  • Purification:
    Immunogen affinity purified.
  • Size:
    100 μg / vial
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Stress-70 protein, mitochondrial(HSPA9) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

HSPA9 (heat shock 70kDa protein 9 (mortalin)), also known as GRP75, mot-2, mthsp75, PBP74, HSPA9B, MORTALIN or MORTALIN, PERINUCLEAR, is a highly conserved member of the HSP70 family of proteins. It functions as a chaperone in the mitochondria, cytoplasm, and centrosome. The HSPA9 gene is mapped to chromosome 5q31.2 based on an alignment of the HSPA9 sequence with the genomic sequence. Knockdown of HSPA9 in erythroid cultures was associated with an increased number of cells in the G0/G1 phase of the cell cycle and accelerated apoptosis. Knockdown of Hspa9 in mouse bone marrow cells, followed by transplantation into wildtype recipients, also resulted in loss of erythroid cell number. Haploinsufficiency for HSPA9 may contribute to abnormal hematopoiesis in myelodysplastic syndromes. This protein plays a role in the control of cell proliferation.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.