Order Entry
Spain
ContactUsLinkComponent
Anti-CD4, CD19, CD20, CD27, CD38, CD45 Mouse Monoclonal Antibody (PerCP-Cy5.5®, PE, redFluor® 710, PE-Cy7®, APC, violetFluor® 450) [clone: OKT4, SJ25C1, 2H7, O323, HIT2, HI30]
Anti-CD4, CD19, CD20, CD27, CD38, CD45 Mouse Monoclonal Antibody (PerCP-Cy5.5®, PE, redFluor® 710, PE-Cy7®, APC, violetFluor® 450) [clone: OKT4, SJ25C1, 2H7, O323, HIT2, HI30]
Supplier:  Tonbo Biosciences
Certificates
Anti-CD4, CD19, CD20, CD27, CD38, CD45 Mouse Monoclonal Antibody (PerCP-Cy5.5®, PE, redFluor® 710, PE-Cy7®, APC, violetFluor® 450) [clone: OKT4, SJ25C1, 2H7, O323, HIT2, HI30]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 91-0100-T100
  • 91-0100-T025
  • Antigen symbol:
  • CD4  
  • CD4  
  • Antigen synonyms:
  • CD38|CD45|CD20|CD27|CD19|CD4
  • CD38|CD45|CD20|CD27|CD19|CD4
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®/PE/redFluor® 710/PE-Cy7®/APC (Allophycocyanin)/violetFluor® 450
  • PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®/PE/redFluor® 710/PE-Cy7®/APC (Allophycocyanin)/violetFluor® 450
  • Clone:
  • OKT4, SJ25C1, 2H7, O323, HIT2, HI30
  • OKT4, SJ25C1, 2H7, O323, HIT2, HI30
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • No
  • No
  • Storage temperature:
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Store undiluted at 2-8 °C, protect from light, do not freeze.

Specifications

Related Information

Frequently Bought Together