Order Entry
Spain
ContactUsLinkComponent
Anti-OXSR1 Antibody Pair
Anti-OXSR1 Antibody Pair
Catalog # ABNOH00009943-PW1
Supplier:  Abnova
Anti-OXSR1 Antibody Pair
Catalog # ABNOH00009943-PW1
Supplier:  Abnova

Specifications

  • Antigen name:
    Oxidative Stress Responsive 1
  • Antigen symbol:
    OXSR1
  • Host:
    Multiple
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    9943
  • Antigen synonyms:
    OXSR 1|KIAA1101|Oxidative stress responsive 1 protein|OSR1|OSR 1|Serine/threonine protein kinase OSR1.|Oxidative stress responsive 1
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

Specifications

About this item

This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-OXSR1 (300 µg); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-OXSR1 (50 µl)

Type:
Antigen: OXSR1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human