Anti-ZNF175 Rabbit Polyclonal Antibody
Catalog # ABNOPAB28682
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger Protein 175
- Antigen symbol:ZNF175
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Cross adsorption:No
- Format:Liquid
- Gene ID:7728
- Antigen synonyms:OTK18
- Storage buffer:PBS, pH 7,2 (40% glycerol, 0,02% sodium azide).
- Sequence:QNQIQPMSHSAFFNKKTLNTESNCEYKDPGKMIRTRPHLASSQKQPQKCCLFTESLKLNLEVNGQNESNDTEQLDDVVGSGQLFSHSSSDACSKNIHTGETFCKGNQCRKVCGHKQSLKQHQ
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids of human ZNF175.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against recombinant ZNF175.
Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Immunofluorescence: 1-4 ?g/ml
Type: Primary
Antigen: ZNF175
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: