Anti-SLC17A7 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: S28-9]
ABNOMAB11528
: Abnova
- Antibody type:Primary
- Antigen name:Solute carrier family 17, member A7
- Antigen symbol:SLC17A7
- Clonality:Monoclonal
- Clone:S28-9
- Conjugation:RPE (R-Phycoerythrin)
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Format:Liquid
- Gene ID:116638
- Antigen synonyms:BNPI|Vglut1
- Amino acid number:493 to 560
- Storage buffer:PBS, pH 7,4 (50% glycerol)
- Storage temperature:Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant fusion protien corresponding to amino acids 493-560 at C-terminus of rat Slc17a7.
- Purification:Protein G Purified
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against recombinant of Slc17a7.
Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000
Type: Primary
Antigen: SLC17A7
Clonality: Monoclonal
Clone: S28-9
Conjugation: RPE (R-Phycoerythrin)
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: