Order Entry
Spain
ContactUsLinkComponent
Anti-IgG Chicken Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-IgG Chicken Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # ABNOPAB29106
Supplier:  Abnova
Anti-IgG Chicken Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # ABNOPAB29106
Supplier:  Abnova

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Monoclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Chicken
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgY
  • Reactivity:
    Mouse
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Form:
    Liquid
  • Storage buffer:
    In PBS, pH 7,2 (0,2% BSA, 0,02% sodium azide)
  • Storage temperature:
    Store at 4 °C and avoid from light. For long term storage, dilute the antibody with sterile glycerol and store it at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Purified mouse IgG (whole protein).
  • Purification:
    Antigen affinity purification
  • Size:
    1 mL
  • Pk:
    1 ml

Specifications

About this item

Chicken polyclonal antibody raised against Mouse IgG. The antibody is conjugated with FITC.

Recommended Dilutions: Immunocytochemistry Immunohistochemistry: 1:250

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Mouse