Order Entry
Spain
ContactUsLinkComponent
Anti-CUL4B Mouse Polyclonal Antibody
Anti-CUL4B Mouse Polyclonal Antibody
Catalog # ABNOH00008450-B01
Supplier:  Abnova
Anti-CUL4B Mouse Polyclonal Antibody
Catalog # ABNOH00008450-B01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cullin 4B
  • Antigen symbol:
    CUL4B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    8450
  • Antigen synonyms:
    CUL-4B|SFM2|MRXS15|MRXSC|MRXHF2
  • Amino acid number:
    1 to 913
  • Storage buffer:
    No additive
  • Storage temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    CUL4B (AAH36216.1, 1 a.a. ~ 913 a.a) full-length human protein.
  • Size:
    50 μl
  • Pk:
    50 µl

Specifications

About this item

Mouse polyclonal antibody raised against a full-length human CUL4B protein.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: CUL4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human