Order Entry
Spain
ContactUsLinkComponent
Anti-USP3 Rabbit Polyclonal Antibody
Anti-USP3 Rabbit Polyclonal Antibody
  ABNOH00009960-D01
 :  Abnova
Anti-USP3 Rabbit Polyclonal Antibody
  ABNOH00009960-D01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Ubiquitin specific peptidase 3
  • Antigen symbol:
    USP3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    9960
  • Antigen synonyms:
    UBP|SIH003
  • Amino acid number:
    1 to 520
  • Storage buffer:
    No additive
  • Storage temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    USP3 (NP_006528.2, 1 a.a. ~ 520 a.a) full-length human protein.
  • Size:
    100 μl
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody raised against a full-length human USP3 protein.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: USP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human