Order Entry
Spain
ContactUsLinkComponent
Anti-ADRBK1 Mouse Monoclonal Antibody [clone: 5D5]
Anti-ADRBK1 Mouse Monoclonal Antibody [clone: 5D5]
Catalog # ABNOMAB2373
Supplier:  Abnova
undefined
Anti-ADRBK1 Mouse Monoclonal Antibody [clone: 5D5]
Catalog # ABNOMAB2373
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Adrenergic Beta, Receptor Kinase 1
  • Antigen symbol:
    ADRBK1
  • Clonality:
    Monoclonal
  • Clone:
    5D5
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Bovine,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Form:
    Tissue Culture Supernatant
  • Gene ID:
    156
  • Antigen synonyms:
    BARK1|BETA-ARK1|beta-adrenergic receptor kinase 1|G-protein coupled receptor kinase 2|GRK2|beta-ARK-1
  • Storage buffer:
    tissue culture supernatant
  • Storage temperature:
    Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to human ADRBK1.
  • Size:
    250 μl
  • Pk:
    250 µl

Specifications

About this item

Mouse monoclonal antibody raised against recombinant ADRBK1.

Recommended Dilutions: Immunohistochemistry: 1:100; Western Blot: 1:100

Type: Primary
Antigen: ADRBK1
Clonality: Monoclonal
Clone: 5D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Bovine, Human, Rat