Anti-SCAND3 Rabbit Polyclonal Antibody
Catalog # ABNOPAB20368
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:SCAN Domain Containing 3
- Antigen symbol:SCAND3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:114821
- Antigen synonyms:SCAND3|ZNF305P2|ZFP38-L|dJ1186N24.3|Buster4|ZNF452
- Storage buffer:PBS, pH 7,5 (40% glycerol, 0,02% sodium azide)
- Sequence:SAFSSEAKLGLSHSQLTEELVASLHTENELDQADKELENTLRAQYEENIETGTDSSDIEENLSVTPKVAEKSPPESRLRFLSCVVCEKECTGVNSCISCDGNIHAICGVPSQHGTEGCGRQITCSLCYETSTMK
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids of human SCAND3.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against recombinant SCAND3.
Recommended Dilutions: Immunohistochemistry: 1:50-1:200
Type: Primary
Antigen: SCAND3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human