Anti-ZIC2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB11336
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger Protein of the cerebellum 2
- Antigen symbol:ZIC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Zebrafish,Frog,Chicken,Chimpanzee,Human,Dog,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:22772
- Antigen synonyms:HPE5|Ku
- Amino acid number:189 to 206
- Storage buffer:20 mM KH2PO4, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 189-206 of mouse Zic2.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of Zic2.
Recommended Dilutions: ELISA: 1:10000-1:50000; Western Blot: 1:500-1:2000
Type: Primary
Antigen: ZIC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Chimpanzee, Dog, Frog, Human, Mouse, Rat, Zebrafish