Order Entry
Spain
ContactUsLinkComponent
Anti-MKI67 Rabbit Polyclonal Antibody
Anti-MKI67 Rabbit Polyclonal Antibody
Catalog # ABNOPAB12128
Supplier:  Abnova
Anti-MKI67 Rabbit Polyclonal Antibody
Catalog # ABNOPAB12128
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Marker of proliferation Ki-67
  • Antigen symbol:
    MKI67
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    17345
  • Antigen synonyms:
    MIB-1|MIB-|KIA|PPP1R105
  • Amino acid number:
    1700 to 1800
  • Storage buffer:
    Tris-citrate/phosphate buffer, pH 7.0-8.0 (0,1% sodium azide)
  • Storage temperature:
    Store at 4 °C. Do not freeze.
  • Immunogen:
    A synthetic peptide corresponding to amino acids 1700-1800 of mouse Mki67.
  • Size:
    100 μl
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of Mki67.

Recommended Dilutions: Immunocytochemistry: 1:50-1:200; Immunofluorescence: 1:50-1:200; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:100

Type: Primary
Antigen: MKI67
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse