Order Entry
Spain
ContactUsLinkComponent
Anti-CDKN2A Rabbit Polyclonal Antibody (DyLight® 549)
Anti-CDKN2A Rabbit Polyclonal Antibody (DyLight® 549)
Catalog # ABNOPAB15345
Supplier:  Abnova
Anti-CDKN2A Rabbit Polyclonal Antibody (DyLight® 549)
Catalog # ABNOPAB15345
Supplier:  Abnova

Specifications

  • Antigen name:
    Cyclin-dependent kinase inhibitor 2A
  • Antigen symbol:
    CDKN2A
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 549
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    12578
  • Antigen synonyms:
    ARF, CDK4I, CDKN2, CMM2, INK4, INK4A, MLM, MTS-1, MTS1, P14, P14ARF, P16, P16-INK4A, P16INK4, P16INK4A, P19, P19ARF, TP16
  • Amino acid number:
    50 to 150
  • Storage buffer:
    50 mM sodium borate
  • Storage temperature:
    Store at 4 °C. Do not freeze.
  • Immunogen:
    A synthetic peptide (conjugated with KLH) corresponding to amino acids 50-150 of mouse Cdkn2a.
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of Cdkn2a.

Recommended Dilutions: Western Blot: 2 ?g/ml; Immunocytochemistry: 1 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1 ?g/ml

Type:
Antigen: CDKN2A
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse