Anti-SERPINB4 Mouse Monoclonal Antibody [clone: 6s40]
Catalog # ABNOMAB8849
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Serpin peptidase inhibitor, clade B (ovalbumin), member 4
- Antigen symbol:SERPINB4
- Clonality:Monoclonal
- Clone:6s40
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:6318
- Antigen synonyms:SCCA-2|Leupin|PI11|Squamous cell carcinoma antigen 2|Peptidase inhibitor 11|Serpin B4|SCCA2|PI-11
- Storage buffer:citrate-Tris-HCl, pH7.0 (0,02% Procl300)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length of human SERPINB4.
- Purification:Affinity purification
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant SERPINB4.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: SERPINB4
Clonality: Monoclonal
Clone: 6s40
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human