Anti-HUWE1 Goat Polyclonal Antibody
Catalog # ABNOPAB17246
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:HECT UBA and WWE Domain Containing 1
- Antigen symbol:HUWE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Epitope:Center
- Cross adsorption:No
- Format:Liquid
- Gene ID:10075
- Antigen synonyms:ARF-BP1, MULE, Ib772, LASU1, UREB1, HECTH9, URE-B1, ARF-BP1, HSPC272, KIAA0312, KIAA1578, E3 ubiquitin-protein ligase HUWE1, ARF-binding protein 1
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-DDLKSNTEYHKYQ
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids at internal region of human HUWE1.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of HUWE1.
Recommended Dilutions: ELISA: 1:128000
Type: Primary
Antigen: HUWE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity: