Order Entry
Spain
ContactUsLinkComponent
Anti-CD56 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MY31]
Anti-CD56 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MY31]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD56 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MY31]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 20-0564-T100
  • 20-0564-T025
  • Antigen symbol:
  • NCAM1
  • NCAM1
  • Antigen name:
  • Neural Cell Adhesion Molecule 1
  • Neural Cell Adhesion Molecule 1
  • Antigen synonyms:
  • NCAM|MSK39|CD56
  • NCAM|MSK39|CD56
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • MY31
  • MY31
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 4684
  • 4684
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • No
  • No
  • Storage temperature:
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Store undiluted at 2-8 °C, protect from light, do not freeze.

Specifications

Related Information

Frequently Bought Together