Anti-CD2 Rat Antibody (PE (Phycoerythrin)) [clone: RM2-5]
The RM2-5 antibody reacts with mouse CD2, an approximately 50 kDa glycoprotein, and a member of the Ig superfamily. CD2, also known as LFA-2, is a receptor for CD48 in the mouse and is expressed on the cell surface of all mouse lymphocytes. CD2 mediates adhesion between cells and is involved in T cell activation.
RM2-5 is reported to block CD2-mediated cell:cell adhesion. Please choose the appropriate format for each application.
Type: Primary
Antigen: CD2
Clonality:
Clone: RM2-5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
- Catalog No:
- 50-0021-U100
- 50-0021-U025
- Antigen symbol:
- CD2
- CD2
- Antigen name:
- CD2 molecule
- CD2 molecule
- Antigen synonyms:
- sheep red blood cell receptor|T-cell surface antigen T11|erythrocyte receptor|rosette receptor|T11|Leu-5|SRBC|LFA-3 receptor|lymphocyte-function antigen-2|LFA-2
- sheep red blood cell receptor|T-cell surface antigen T11|erythrocyte receptor|rosette receptor|T11|Leu-5|SRBC|LFA-3 receptor|lymphocyte-function antigen-2|LFA-2
- Antibody type:
- Primary
- Primary
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- RM2-5
- RM2-5
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 12481
- 12481
- Isotype:
- IgG2b
- IgG2b
- Flow cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Storage temperature:
- Store undiluted at 2...8 °C, protect from light, do not freeze
- Store undiluted at 2...8 °C, protect from light, do not freeze
- Tested applications:
- Flow cytometry
- Flow cytometry
Frequently Bought Together