Anti-CD3E Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: OKT3]
BLEG317332
- Antibody type:Primary
- Antigen name:CD3 molecule epsilon
- Antigen symbol:CD3E
- Clonality:Monoclonal
- Clone:OKT3
- Conjugation:Brilliant Violet® 510
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Gene ID:916
- Antigen synonyms:CD3e antigen|TCRE|epsilon subunit of T3|epsilon polypeptide TiT3 complex|T3E|FLJ18683|CD3-epsilon|T-cell surface glycoprotein CD3 epsilon chain|T-cell antigen receptor complex|T-cell surface antigen T3/Leu-4 epsilon chain
- Storage temperature:The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:Ambient RT
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 510® under optimal conditions. The solution is free of unconjugated Brilliant Violet 510® and unconjugated antibody.
- Size:100 Tests
- Pk:100 Tests
Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: OKT3
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: