Anti-Heat Shock Factor 1 Rabbit Polyclonal Antibody
Catalog # USBIH1832-25-200
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Heat Shock Factor 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rabbit,Monkey,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide, 50% glycerol.
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Recombinant human HSF-1 expressed in E.coli.
- Purification:purified by Protein A chromatography
- Size:200 μg
- Pk:200 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: Heat Shock Factor 1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human, Monkey, Mouse, Rabbit, Rat,