Anti-MTDH Mouse Monoclonal Antibody [clone: 2F11]
Catalog # ORIGTA803710
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:MTDH
- Clonality:Monoclonal
- Clone:2F11
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:70 to 582
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 70-582 of human MTDH (NP_848927)produced in SF9 cell.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: MTDH
Clonality: Monoclonal
Clone: 2F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human