Anti-TRIM63 Rabbit Polyclonal Antibody
100 μl
Catalog # ORIGTA322135
Supplier: OriGene
Jump to:
Product Details & Documents
Recommended Dilutions: ELISA: 1:2000-10000; Western Blot: 1:1000-5000; Immunochemistry: 1:25-100
Type: Primary
Antigen: TRIM63
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
TA322135
Datasheet
Specifications
- Size:100 μl
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by antigen affinity chromatography
- Antibody type:Primary
- Form:Liquid
- Amino acid number:2 to 14
- Isotype:IgG
- Storage temperature:–20 °C
- Host:Rabbit
- Cross adsorption:No
- Molecular weight:40 kDa
- Shipping temperature:–20 °C
- Cat. No.:ORIGTA322135
- Reactivity:Human|Rat|Mouse
- Immunogen:Synthetic peptide peptide corresponding to a region derived from 2-14 amino acids of human tripartite motif containing 63, E3 ubiquitin protein ligase
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:TRIM63
- Storage buffer:PBS pH 7,3, 0,05% NaN3, 50% glycerol
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



